tert-Butyl-4-oxoazepane-1-carboxylate 95%

Supplier: Ambeed
Warning

A244693-100G A244693-10G A244693-250MG A244693-1G A244693-5G A244693-25G
103811-596EA 6 USD
103811-596 103851-430 77013-844 77013-846 77013-848 77090-638 77090-640 77304-330
tert-Butyl-4-oxoazepane-1-carboxylate 95%
tert-Butyl-4-oxoazepane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
Boiling Pt: 333.3 °C (760 mm Hg)
Melting Pt: 39…43 °C
Flash Pt: 140.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788435
CAS Number: 188975-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR