1-Boc-2-trifluoromethyl-4-piperidone 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-oxo-2-(trifluoromethyl)piperidine-1-carboxylate

A210794-100MG A210794-10G A210794-250MG A210794-50MG A210794-1G A210794-5G
103818-044EA 178 USD
103818-044 103848-736 77030-486 77030-488 77030-490 77030-492 77087-122 77303-148
1-Boc-2-trifluoromethyl-4-piperidone 98%
1-Boc-2-trifluoromethyl-4-piperidone

Precaución: for Research Use Only

Formula: C₁₁H₁₆F₃NO₃
MW: 267.248 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07367733
CAS Number: 1245648-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR