tert-Butyl 4-methylpiperazine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A231223-1G
A231223-5G
A231223-100G
A231223-10G
A231223-25G
103810-764EA
5
USD
103810-764
103850-512
77089-392
77089-394
77165-536
77165-538
77303-906
tert-Butyl 4-methylpiperazine-1-carboxylate 97%
tert-Butyl 4-methylpiperazine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Boiling Pt: 254.8 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD03840525 CAS Number: 53788-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...