1-Boc-4-iodopiperidine 98%

Supplier: Ambeed
Warning

Synonyms: 1-BOC-4-iodo-piperidine, tert-Butyl 4-iodopiperidine-1-carboxylate

A663285-1G A663285-5G A663285-100G A663285-10G A663285-250MG A663285-500G
103840-452EA 15 USD
103840-452 76844-640 76844-642 77170-480 77170-482 77170-484 77614-046
1-Boc-4-iodopiperidine 98%
1-Boc-4-iodopiperidine

Precaución: for Research Use Only

Formula: C₁₀H₁₈INO₂
MW: 311.16 g/mol
Boiling Pt: 318.8 °C (760 mm Hg)
Melting Pt: 43…47 °C
Flash Pt: 146.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115041
CAS Number: 301673-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR