tert-Butyl 4-iodobenzoate 95%

Supplier: Ambeed
Warning

A187295-10G A187295-250MG A187295-1G A187295-25G A187295-5G
103816-000EA 35 USD
103816-000 103846-590 76985-982 76985-984 77302-070 77302-072 77302-074
tert-Butyl 4-iodobenzoate 95%
tert-Butyl 4-iodobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃IO₂
Boiling Pt: 310.2±25.0 °C (760 mm Hg)
Flash Pt: 141.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11501908
CAS Number: 120363-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR