tert-Butyl 4-iodo-1H-pyrazole-1-carboxylate 97%

Supplier: Ambeed
Warning

A685089-10G A685089-250MG A685089-1G A685089-25G A685089-5G
103840-554EA 22 USD
103840-554 76790-992 76790-994 76844-764 76844-766 76844-768
tert-Butyl 4-iodo-1H-pyrazole-1-carboxylate 97%
tert-Butyl 4-iodo-1H-pyrazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁IN₂O₂
Boiling Pt: 312 °C (760 mm Hg)
Melting Pt: 67…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663855
CAS Number: 121669-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR