1-Boc-4-hydroxypiperidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate
A214139-100G
A214139-10G
A214139-500G
A214139-5G
A214139-1000G
A214139-25G
103809-494EA
203
USD
103809-494
76770-632
76770-634
76770-636
76770-638
77087-578
77087-580
1-Boc-4-hydroxypiperidine 98%
1-Boc-4-hydroxypiperidine
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₃ MW: 201.27 Melting Pt: 61…65 °C Flash Pt: 130.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075174 CAS Number: 109384-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...