tert-Butyl-4-hydroxyphenethylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-4-hydroxyphenethylamine, tert-Butyl 4-hydroxyphenethylcarbamate, N-(tert-Butoxycarbonyl)-4-hydroxyphenethylamine, N-Boc-tyramine, N-(tert-Butoxycarbonyl)tyramine

A351932-100G A351932-10G A351932-250MG A351932-1G A351932-25G A351932-5G
103826-058EA 9 USD
103826-058 103837-054 76724-474 76724-476 76724-478 76724-480 76840-166 76840-168 76840-170
tert-Butyl-4-hydroxyphenethylcarbamate 97%
tert-Butyl-4-hydroxyphenethylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
Melting Pt: 71…75 °C
Flash Pt: 193.1 °C
Storage Temperature: Ambient
MDL Number: MFCD05864730
CAS Number: 64318-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR