tert-Butyl 4-Formylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-formylbenzoate, 4-Formylbenzoic acid tert-Butyl ester, 4-(tert-Butoxycarbonyl)benzaldehyde

A129714-100MG A129714-250MG A129714-25G A129714-1G A129714-5G A129714-10G
103801-314EA 10 USD
103801-314 77285-262 77285-264 77285-266 77313-706 77313-708 77566-518
tert-Butyl 4-Formylbenzoate 97%
tert-Butyl 4-Formylbenzoate

Precaución: For research use only.

Formula: C₁₂H₁₄O₃
Boiling Pt: 310.7 °C (760 mm Hg)
Flash Pt: 134.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01111994
CAS Number: 65874-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR