tert-Butyl 4-formyl-1H-pyrazole-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-formylpyrazole-1-carboxylate

A114821-100MG A114821-250MG A114821-1G A114821-5G
103798-482EA 25 USD
103798-482 103824-324 76971-106 76971-108 77297-332 77297-334
tert-Butyl 4-formyl-1H-pyrazole-1-carboxylate 97%
tert-Butyl 4-formyl-1H-pyrazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₃
MW: 196.203 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19707636
CAS Number: 821767-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR