tert-Butyl 4-cyano-5,6-dihydropyridine-1(2H)-carboxylate 97%

Supplier: Ambeed
Warning

A387431-100MG A387431-250MG
103826-826EA 352 USD
103826-826 76990-082 76990-084
tert-Butyl 4-cyano-5,6-dihydropyridine-1(2H)-carboxylate 97%
tert-Butyl 4-cyano-5,6-dihydropyridine-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₂
MW: 208.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22665818
CAS Number: 873551-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR