1-Boc-4-chloropiperidine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-chloropiperidine-1-carboxylate

A181953-10G A181953-250MG A181953-1G A181953-5G A181953-25G
103807-672EA 14 USD
103807-672 103846-042 77012-022 77012-024 77083-570 77083-572 77301-762
1-Boc-4-chloropiperidine 97%
1-Boc-4-chloropiperidine

Precaución: for Research Use Only

Formula: C₁₀H₁₈ClNO₂
Boiling Pt: 289.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04115040
CAS Number: 154874-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR