tert-Butyl-4-carbamoylpiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-carbamoylpiperidine-1-carboxylate, 1-Boc-4-piperidinecarboxamide, tert-Butyl 4-Carbamoyl-1-piperidinecarboxylate, 1-(tert-Butoxycarbonyl)-4-piperidinecarboxamide, 4-Carbamoyl-1-piperidinecarboxylic acid tert-Butyl ester

A194749-100G A194749-10G A194749-500G A194749-25G A194749-1G
103816-690EA 6 USD
103816-690 103847-302 76800-124 76800-126 76800-128 77085-226 77302-434
tert-Butyl-4-carbamoylpiperidine-1-carboxylate 97%
tert-Butyl-4-carbamoylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₃
MW: 228.2900 g/mol
Boiling Pt: 384.4 °C (760 mm Hg)
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD02180953
CAS Number: 91419-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR