tert-Butyl-4-carbamothioylpiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 4-Carbamothioylpiperidine, N1-BOC protected, tert-Butyl 4-carbamothioylpiperidine-1-carboxylate

A209255-100G A209255-10G A209255-500G A209255-25G A209255-5G A209255-1G
103817-926EA 10 USD
103817-926 103848-614 76785-638 76785-640 76785-642 77086-954 77086-956 77303-094
tert-Butyl-4-carbamothioylpiperidine-1-carboxylate 95%
tert-Butyl-4-carbamothioylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂S
MW: 244.3500 g/mol
Boiling Pt: 356 °C (760 mm Hg)
Melting Pt: 130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02180954
CAS Number: 214834-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR