tert-Butyl 4-bromobutanoate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-bromobutyrate

A108512-10G A108512-25G A108512-1G A108512-5G A108512-100G A108512-500G
103797-358EA 7 USD
103797-358 103823-138 76780-968 76780-970 77309-982 77309-984 77599-570 77666-576
tert-Butyl 4-bromobutanoate 97%
tert-Butyl 4-bromobutanoate

Precaución: for Research Use Only

Formula: C₈H₁₅BrO₂
MW: 223.107 g/mol
Boiling Pt: 225.9 °C (760 mm Hg)
Flash Pt: 117.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179416
CAS Number: 110661-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR