tert-Butyl 4-bromobenzylcarbamate 98%

Supplier: Ambeed
Warning

A349997-10G A349997-250MG A349997-1G A349997-25G A349997-5G A349997-100G
103826-032EA 8 USD
103826-032 103837-026 76788-380 76788-382 76840-126 76840-128 76840-130 77668-600
tert-Butyl 4-bromobenzylcarbamate 98%
tert-Butyl 4-bromobenzylcarbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂
Boiling Pt: 370.3 °C (760 mm Hg)
Melting Pt: 86…88 °C
Flash Pt: 177.8 °C
Storage Temperature: Ambient
MDL Number: MFCD08703140
CAS Number: 68819-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR