tert-Butyl 4-bromo-5,6-dihydropyridine-1(2H)-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-bromo-1,2,3,6-tetrahydropyridine-1-carboxylate, tert-Butyl 4-bromo-3,6-dihydro-2H-pyridine-1-carboxylate

A135591-250MG A135591-1G
103802-180EA 10 USD
103802-180 77010-118 77315-024
tert-Butyl 4-bromo-5,6-dihydropyridine-1(2H)-carboxylate 95%
tert-Butyl 4-bromo-5,6-dihydropyridine-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆BrNO₂
MW: 262.144 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181893
CAS Number: 159503-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR