4-Amino-1-boc-piperidine 98%

Supplier: Ambeed
Warning

A143361-100G A143361-10G A143361-1G A143361-500G A143361-25G A143361-5G
76767-902EA 66.84 USD
76767-902 76767-904 76767-906 76767-908 77316-530 77316-532
4-Amino-1-boc-piperidine 98%
4-Amino-1-Boc-piperidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Flash Pt: 121.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01076201
CAS Number: 87120-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR