4-[2-(Boc-amino)ethyl]aniline 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-aminophenethylcarbamate, 4-(2-Aminoethyl)aniline, 4-BOC protected

A113213-100G A113213-100MG A113213-10G A113213-250MG A113213-25G A113213-1G A113213-5G
103798-192EA 8 USD
103798-192 103824-030 77148-816 77148-818 77148-820 77148-822 77148-824 77310-856 77310-858
4-[2-(Boc-amino)ethyl]aniline 97%
4-[2-(Boc-amino)ethyl]aniline

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₂
MW: 236.315 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05663961
CAS Number: 94838-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR