tert-Butyl 4-aminoisoindoline-2-carboxylate 97%

Supplier: Ambeed
Warning

A185372-100MG A185372-250MG A185372-50MG
103815-846EA 176 USD
103815-846 103846-410 76825-078 76825-080 76825-082
tert-Butyl 4-aminoisoindoline-2-carboxylate 97%
tert-Butyl 4-aminoisoindoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566550
CAS Number: 871013-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR