tert-Butyl-4-aminobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 4-aminobenzoate
A235339-100G
A235339-10G
A235339-500G
A235339-25G
A235339-5G
103850-772EA
7
USD
103850-772
76801-174
76801-176
76801-178
77089-782
77089-784
tert-Butyl-4-aminobenzoate 98%
tert-Butyl-4-aminobenzoate
Precaución: for Research Use Only
Formula:
C₁₁H₁₅NO₂ MW: 193.2423 g/mol Boiling Pt: 322.4 °C (760 mm Hg) Melting Pt: 108…110 °C Flash Pt: 173.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00665790 CAS Number: 18144-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...