tert-Butyl 4-amino-4-(aminomethyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A241838-100MG A241838-250MG
103811-434EA 109 USD
103811-434 103851-240 76786-558 76786-560
tert-Butyl 4-amino-4-(aminomethyl)piperidine-1-carboxylate 97%
tert-Butyl 4-amino-4-(aminomethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₃N₃O₂
MW: 229.32 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08234568
CAS Number: 871115-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR