tert-Butyl 4-amino-3-fluoropiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A309749-1G A309749-5G A309749-100MG A309749-250MG A309749-25G
103814-598EA 75 USD
103814-598 103835-780 76838-550 76838-552 76977-884 77672-576 77881-863
tert-Butyl 4-amino-3-fluoropiperidine-1-carboxylate 95%
tert-Butyl 4-amino-3-fluoropiperidine-1-carboxylate

Precaución: For research use only

Formula: C₁₀H₁₉FN₂O₂
MW: 218.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566556
CAS Number: 934536-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR