tert-Butyl 4-amino-3,3-difluoropyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A230176-50MG A230176-100MG A230176-250MG
103792-460EA 56 USD
103792-460 103799-514 77303-864 77892-383 77892-384
tert-Butyl 4-amino-3,3-difluoropyrrolidine-1-carboxylate 95%
tert-Butyl 4-amino-3,3-difluoropyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₆F₂N₂O₂
Boiling Pt: 261.7±40.0 °C (760 mm Hg)
Flash Pt: 112.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD23105898
CAS Number: 1408074-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR