tert-Butyl 4-amino-3,3-difluoropiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A164117-100MG A164117-250MG A164117-1G A164117-5G
103805-818EA 84 USD
103805-818 103844-082 76872-132 76872-134 77320-226 77320-228
tert-Butyl 4-amino-3,3-difluoropiperidine-1-carboxylate 95%
tert-Butyl 4-amino-3,3-difluoropiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈F₂N₂O₂
MW: 236.259 g/mol
Boiling Pt: 281.5±40.0 °C (760 mm Hg)
Flash Pt: 124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16250242
CAS Number: 1255666-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR