tert-Butyl 4-amino-2-oxo-5,6-dihydropyridine-1(2H)-carboxylate 95%

Supplier: Ambeed
Warning

A654155-1G A654155-250MG
103840-410EA 660 USD
103840-410 76844-588 76992-422
tert-Butyl 4-amino-2-oxo-5,6-dihydropyridine-1(2H)-carboxylate 95%
tert-Butyl 4-amino-2-oxo-5,6-dihydropyridine-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₃
MW: 212.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18207247
CAS Number: 1333319-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR