tert-Butyl-4,7-diazaspiro[2.5]octane-7-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4,7-diazaspiro[2.5]octane-7-carboxylate, 4,7-Diaza-spiro(2.5)octane-7-carboxylic acid tert-butyl ester

A130297-250MG A130297-1G A130297-100MG
103801-408EA 53 USD
103801-408 76972-032 77313-840 77670-844
tert-Butyl-4,7-diazaspiro[2.5]octane-7-carboxylate 95%
tert-Butyl-4,7-diazaspiro[2.5]octane-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MW: 212.2887 g/mol
Boiling Pt: 301.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08685931
CAS Number: 886766-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR