tert-Butyl 4-(pyrimidin-5-yl)piperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

A225813-250MG A225813-1G A225813-5G
103819-200EA 36 USD
103819-200 103850-102 76826-304 77303-750 77303-752
tert-Butyl 4-(pyrimidin-5-yl)piperazine-1-carboxylate 98%
tert-Butyl 4-(pyrimidin-5-yl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₄O₂
MW: 264.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11872512
CAS Number: 634468-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR