tert-Butyl-4-(iodomethyl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(iodomethyl)piperidine-1-carboxylate

A275595-100G A275595-100MG A275595-10G A275595-250MG A275595-1G A275595-5G
103813-270EA 14 USD
103813-270 103853-194 76988-392 76988-394 76988-396 76988-398 77093-048 77093-050
tert-Butyl-4-(iodomethyl)piperidine-1-carboxylate 95%
tert-Butyl-4-(iodomethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀INO₂
Boiling Pt: 336.6 °C (760 mm Hg)
Flash Pt: 157.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08437653
CAS Number: 145508-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR