tert-Butyl-4-(hydroxymethyl)benzylcarbamate 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Aminomethyl)benzyl alcohol, N-BOC protected, 4-(Boc-aminomethyl-phenyl)-methanol, tert-Butyl 4-(hydroxymethyl)benzylcarbamate

A101402-100MG A101402-250MG A101402-1G A101402-100G A101402-25G
103795-958EA 20 USD
103795-958 103821-642 77008-332 77008-334 77297-090 77682-935 77682-936
tert-Butyl-4-(hydroxymethyl)benzylcarbamate 95%
tert-Butyl-4-(hydroxymethyl)benzylcarbamate

Precaución: For research use only.

Formula: C₁₃H₁₉NO₃
MW: 237.3 g/mol
Boiling Pt: 397.2 °C (760 mm Hg)
Melting Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06411553
CAS Number: 123986-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR