tert-Butyl-4-(bromomethyl)benzylcarbamate 97%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 4-(bromomethyl)benzylcarbamate, N-BOC-4-(Bromomethyl)benzylamine

A193494-100MG A193494-10G A193494-250MG
103816-548EA 47 USD
103816-548 103847-164 76974-930 77611-848 77611-850
tert-Butyl-4-(bromomethyl)benzylcarbamate 97%
tert-Butyl-4-(bromomethyl)benzylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₂
MW: 300.1900 g/mol
Flash Pt: 191.071 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11974929
CAS Number: 187283-17-6
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR