4-(6-Amino-3-pyridyl)-1-boc-piperazine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Boc-4-(6-amino-3-pyridyl)piperazine, tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate, tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate, 1-tert-Butoxycarbonyl-4-(6-amino-3-pyridyl)piperazine, 4-(6-Amino-3-pyridyl)piperazine-1-carboxylic acid tert-Butyl ester
A147365-100G
A147365-10G
A147365-1G
A147365-25G
A147365-5G
A147365-500G
103803-764EA
5
USD
103803-764
103841-864
76812-034
76812-036
77317-236
77317-238
77317-240
77883-165
4-(6-Amino-3-pyridyl)-1-boc-piperazine 98%
4-(6-Amino-3-pyridyl)-1-boc-piperazine
Precaución: for Research Use Only
Formula:
C₁₄H₂₂N₂O₄ Melting Pt: 130…132 °C Flash Pt: 228.411 °C Storage Temperature: Ambient |
MDL Number:
MFCD11594962 CAS Number: 571188-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...