4-(5-Amino-1H-pyrazol-3-yl)piperidine-1-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(5-amino-1H-pyrazol-3-yl)piperidine-1-carboxylate

A152203-100MG A152203-250MG A152203-5G
103804-360EA 42 USD
103804-360 103842-522 76984-676 76984-678 77299-936
4-(5-Amino-1H-pyrazol-3-yl)piperidine-1-carboxylic acid tert-butyl ester 95%
4-(5-Amino-1H-pyrazol-3-yl)piperidine-1-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₄O₂
MW: 266.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16618570
CAS Number: 1169563-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR