1-(1-Boc-piperidino)-4-bromo-1H-pyrazole 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(4-bromo-1H-pyrazol-1-yl)piperidine-1-carboxylate, 1-(4-Boc-1-piperidinyl)-4-bromopyrazole

A249791-1G A249791-5G A249791-100G A249791-250MG A249791-25G
103811-890EA 13 USD
103811-890 103851-728 77091-074 77091-076 77288-980 77288-982 77304-500
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole 95%
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₃H₂₀BrN₃O₂
MW: 330.23 g/mol
Boiling Pt: 411.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09831980
CAS Number: 877399-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR