tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(4-aminophenyl)piperazine, tert-Butyl 4-(4-aminophenyl)piperazine-1-carboxylate

A396431-1G A396431-25G A396431-5G A396431-10G A396431-250MG
103826-980EA 6 USD
103826-980 103838-020 76841-446 76841-448 76841-450 77016-258 77016-260
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate 98%
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₃N₃O₂
Boiling Pt: 441.1 °C (760 mm Hg)
Flash Pt: 220.593 °C
Storage Temperature: Ambient
MDL Number: MFCD04115065
CAS Number: 170911-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR