tert-Butyl-4-(4-amino-1H-pyrazol-1-yl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(4-amino-1H-pyrazol-1-yl)piperidine-1-carboxylate

A850304-1G A850304-25G A850304-5G A850304-10G A850304-250MG
103841-234EA 104 USD
103841-234 76845-480 76845-482 76845-484 77159-504 77159-506
tert-Butyl-4-(4-amino-1H-pyrazol-1-yl)piperidine-1-carboxylate 97%
tert-Butyl-4-(4-amino-1H-pyrazol-1-yl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₄O₂
MW: 266.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10687120
CAS Number: 1029413-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR