4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylcarbamate, 4-(N-Boc-aminomethyl)phenylboronic acid pinacol ester, [4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzyl]-carbamic acid tert-butyl ester

A114471-100MG A114471-10G A114471-250MG A114471-25G A114471-1G A114471-5G
103798-414EA 14 USD
103798-414 103824-252 77284-390 77284-392 77284-394 77284-396 77311-060 77311-062
4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester 95%
4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₈BNO₄
MW: 333.235 g/mol
Boiling Pt: 452.0±38.0 °C (760 mm Hg)
Flash Pt: 227.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06409942
CAS Number: 330794-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR