tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-BOC-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate, 1(2H)-Pyridinecarboxylic acid, 3,6-dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 1,1-dimethylethyl ester, N-Boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, 2-Methyl-2-propanyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-1(2H)-pyridinecarboxylate
A130704-1000G
A130704-100G
A130704-10G
A130704-500G
A130704-1G
A130704-25G
A130704-5G
103801-466EA
8
USD
103801-466
77094-960
77094-962
77094-964
77094-966
77298-426
77313-922
77313-924
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 97%
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate
Precaución: for Research Use Only
Formula:
C₁₆H₂₈BNO₄ MW: 309.21 g/mol Boiling Pt: 348 °C (760 mm Hg) Melting Pt: 100…114 °C Flash Pt: 164.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840345 CAS Number: 286961-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...