tert-Butyl 4-(4-(hydroxymethyl)phenoxy)piperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A149937-100MG A149937-250MG A149937-1G
103804-102EA 50 USD
103804-102 103842-230 76647-044 76647-046 77299-762
tert-Butyl 4-(4-(hydroxymethyl)phenoxy)piperidine-1-carboxylate 95%
tert-Butyl 4-(4-(hydroxymethyl)phenoxy)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₅NO₄
MW: 307.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31657114
CAS Number: 321337-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR