4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine, N1-BOC protected 95%

Supplier: Ambeed
Warning

Synonyms: 4-[4-(tert-Butoxycarbonyl)piperazin-1-yl]benzeneboronic acid, pinacol ester, tert-Butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine-1-carboxylate, tert-Butyl 4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazine-1-carboxylate

A319256-10G A319256-250MG A319256-1G A319256-25G
103825-196EA 9 USD
103825-196 103836-128 77290-268 77290-270 77306-198 77306-200
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine, N1-BOC protected 95%
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine, N1-BOC protected

Precaución: for Research Use Only

Formula: C₂₁H₃₃BN₂O₄
MW: 388.309 g/mol
Boiling Pt: 501.7 °C (760 mm Hg)
Melting Pt: 157…159 °C
Flash Pt: 257.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06411544
CAS Number: 470478-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR