tert-Butyl-4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(3-hydroxypropyl)piperidine-1-carboxylate, 4-(3-Hydroxyprop-1-yl)piperidine, N-BOC protected, tert-Butyl 4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate

A165055-250MG A165055-1G A165055-25G
103805-938EA 15 USD
103805-938 103844-196 76720-272 77320-388 77320-390
tert-Butyl-4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate 98%
tert-Butyl-4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₅NO₃
MW: 243.35 g/mol
Boiling Pt: 339.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02677712
CAS Number: 156185-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR