tert-Butyl 4-(3-aminophenyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A209630-100MG A209630-250MG A209630-1G
103817-950EA 150 USD
103817-950 77287-928 77287-930 77303-102
tert-Butyl 4-(3-aminophenyl)piperidine-1-carboxylate 97%
tert-Butyl 4-(3-aminophenyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂O₂
MW: 276.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701248
CAS Number: 387827-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR