N-Boc-4-piperidineacetaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(N-BOC-4-piperidinyl)acetaldehyde, tert-Butyl 4-(2-oxoethyl)piperidine-1-carboxylate
A121901-100MG
A121901-10G
A121901-250MG
A121901-25G
A121901-1G
A121901-5G
103800-154EA
42
USD
103800-154
77284-850
77284-852
77284-854
77284-856
77312-284
77312-286
N-Boc-4-piperidineacetaldehyde 98%
N-Boc-4-piperidineacetaldehyde
Precaución: for Research Use Only
Formula:
C₁₂H₂₁NO₃ MW: 227.31 g/mol Boiling Pt: 318.1 °C (760 mm Hg) Melting Pt: 38…42 °C Flash Pt: 110 °C Storage Temperature: Freezer |
MDL Number:
MFCD03425258 CAS Number: 142374-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...