tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate 97%

Supplier: Ambeed
Warning

A108709-100G A108709-10G A108709-250MG A108709-25G A108709-500G A108709-1G A108709-5G
103797-382EA 10 USD
103797-382 103823-166 76995-072 76995-074 76995-076 76995-078 76995-080 77283-770 77283-772
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate 97%
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₄
Boiling Pt: 354 °C (760 mm Hg)
Flash Pt: 168 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12912655
CAS Number: 135716-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR