tert-Butyl-4-(2-ethoxy-2-oxoethyl)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(2-ethoxy-2-oxoethyl)piperidine-1-carboxylate

A371806-100G A371806-10G A371806-500G
103826-494EA 8 USD
103826-494 103837-512 76724-780 77572-112 77668-734
tert-Butyl-4-(2-ethoxy-2-oxoethyl)piperidine-1-carboxylate 98%
tert-Butyl-4-(2-ethoxy-2-oxoethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₄
MW: 271.36 g/mol
Boiling Pt: 339.2±15.0 °C (760 mm Hg)
Flash Pt: 158.9 °C
Storage Temperature: Ambient
MDL Number: MFCD08275143
CAS Number: 135716-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR