4-(Cyanoacetyl)piperidine, N-BOC protected 97%

Supplier: Ambeed
Warning

A237545-1G A237545-100MG A237545-250MG
103811-158EA 21 USD
103811-158 103850-936 77090-026 77193-250 77193-252
4-(Cyanoacetyl)piperidine, N-BOC protected 97%
4-(Cyanoacetyl)piperidine, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD08059465
CAS Number: 660406-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR