tert-Butyl-4-(2-chloropyrimidin-4-yl)piperazine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 4-(2-chloropyrimidin-4-yl)piperazine-1-carboxylate
A101045-250MG
A101045-1G
A101045-5G
A101045-25G
103795-778EA
21
USD
103795-778
103821-468
76780-444
77296-966
77296-968
77599-478
tert-Butyl-4-(2-chloropyrimidin-4-yl)piperazine-1-carboxylate 97%
tert-Butyl-4-(2-chloropyrimidin-4-yl)piperazine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₁₉ClN₄O₂ MW: 298.77 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03407443 CAS Number: 221050-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...