tert-Butyl 4-(2-bromoethyl)piperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A116534-250MG A116534-1G A116534-100MG
103798-746EA 35 USD
103798-746 103824-612 77284-556 77311-420 77747-847
tert-Butyl 4-(2-bromoethyl)piperazine-1-carboxylate 97%
tert-Butyl 4-(2-bromoethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁BrN₂O₂
MW: 293.21 g/mol
Flash Pt: 154.3 °C
Storage Temperature: Freezer
MDL Number: MFCD07367890
CAS Number: 655225-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR