tert-Butyl-4-(2-bromoacetyl)piperidine-1-carboxylate 95%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 4-(2-bromoacetyl)piperidine-1-carboxylate

A119030-250MG A119030-25G A119030-1G A119030-5G A119030-10G
103799-150EA 16 USD
103799-150 76810-360 76810-362 77297-654 77311-822 77566-196
tert-Butyl-4-(2-bromoacetyl)piperidine-1-carboxylate 95%
tert-Butyl-4-(2-bromoacetyl)piperidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₂H₂₀BrNO₃
MW: 306.21 g/mol
Boiling Pt: 368 °C (760 mm Hg)
Flash Pt: 176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11975555
CAS Number: 301221-79-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR