tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-BOC-4-(2-aminoethyl)piperazine, 4-(2-Aminoethyl)-1-boc-piperazine, tert-Butyl 4-(2-aminoethyl)piperazine-1-carboxylate
A113040-10G
A113040-1G
A113040-5G
A113040-25G
A113040-500G
103798-152EA
7
USD
103798-152
103823-988
76796-940
77310-824
77310-826
77549-194
77666-644
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate 97%
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₂₃N₃O₂ MW: 229.32 g/mol Boiling Pt: 319.4 °C (760 mm Hg) Melting Pt: 36…41 °C Flash Pt: 198 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683049 CAS Number: 192130-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...